missing translation for 'onlineSavingsMsg'
Learn More

BD Cytofix™ Fixation Buffer

Product Code. 10076765
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 10076765

Brand: BD 554655

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Can be used to fix unstained cells for subsequent immunofluorescent staining of intracellular cytokines. Can also be used to fix cells after immunofluorescent staining in order to preserve the light-scattering signals and fluorescent intensities of cells for analysis at a later time.

  • Intended to preserve human and rodent lymphoid cells for subsequent immunofluorescent staining of intracellular cytokines
  • Used to preserve light-scattering characteristics and fluorescence intensities of human and rodent hematopoietic cells that have been stained by immunofluorescence for subsequent flow cytometric analysis 
TRUSTED_SUSTAINABILITY

Specifications

Formulation Neutral pH Buffered Saline (PBS) with 4.21% Formaldehyde (w/w)
For Use With (Application) Can be used to fix unstained cells for subsequent immunofluorescent staining of intracellular cytokines
Content And Storage 4°C
Product Type Fixation Buffer
Quantity 100 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.