missing translation for 'onlineSavingsMsg'
Learn More

BD Cytofix/Cytoperm™ Fixation/Permeabilization Solution Kit with BD GolgiStop™

Product Code. 15867858 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
250 Tests
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15867858 250 Tests 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15867858 Supplier BD Supplier No. 554715
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

  • Fixation/Permeabilization solution (125mL)
  • BD Perm/Wash™ Buffer, 10x concentrate containing Fetal Bovine Serum (FBS) and saponin (100mL)
  • Protein transport inhibitor BD GolgiStop™ (containing Monensin)

Provides GolgiStop protein transport inhibitor containing monensin

The ex vivo addition of GolgiStop to in vitro- or in vivo -stimulated cells blocks their intracellular transport processes. This results in the accumulation of most cytokine proteins in the Golgi complex and thereby enhances cytokine staining signals.

  • GolgiStop reagent is provided for treating up to 1L of cell culture at a cell density of up to 2 × 106 cells/mL

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 4°C
Includes 125 mL fixation/permeabilization solution, perm/Wash™ buffer, fetal bovine serum (FBS) 10X and 100 mL saponin, protein transport inhibitor BD GolgiStop™
No. of Tests 250
Quantity 250 Tests
Regulatory Status RUO
Product Type Fixation/Permeabilization Reagent
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • BD Cytofix/Cytoperm Plus Fixation/Permeabilization Kit with Golgistop
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1A
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol, monensin mononatriumsalz, formaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.