missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Cytofix/Cytoperm™ Fixation/Permeabilization Solution Kit with BD GolgiStop™
Click to view available options
Quantity:
250 Tests
Unit Size:
250 tests
Description
Includes
- Fixation/Permeabilization solution (125mL)
- BD Perm/Wash™ Buffer, 10x concentrate containing Fetal Bovine Serum (FBS) and saponin (100mL)
- Protein transport inhibitor BD GolgiStop™ (containing Monensin)
The ex vivo addition of GolgiStop to in vitro- or in vivo -stimulated cells blocks their intracellular transport processes. This results in the accumulation of most cytokine proteins in the Golgi complex and thereby enhances cytokine staining signals.
- GolgiStop reagent is provided for treating up to 1L of cell culture at a cell density of up to 2 × 106 cells/mL
Specifications
Specifications
| Content And Storage | 4°C |
| Includes | 125 mL fixation/permeabilization solution, perm/Wash™ buffer, fetal bovine serum (FBS) 10X and 100 mL saponin, protein transport inhibitor BD GolgiStop™ |
| No. of Tests | 250 |
| Quantity | 250 Tests |
| Regulatory Status | RUO |
| Product Type | Fixation/Permeabilization Reagent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction