missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD CD1d:Ig Recombinant Fusion Protein, Mouse DimerX
Click to view available options
Quantity:
0.25 mg
Unit Size:
Each
Description
The CD1d:Ig fusion protein consists of extracellular major histocompatibility complex (MHC) class I CD1d domains that are fused with VH regions of mouse IgG1. Like MHC class I molecules, the CD1d noncovalently associates with β2 Microglobulin ( β2M). Recombinant CD1d molecules are useful for studying Natural Killer T (NKT)-cell function by immunofluorescent staining and flow cytometric analysis of antigen-specific NKT cells.
- Format: Purified
- Clone: DimerX/ CD1d
- Isotype: Mouse IgG1, λ
- Concentration: 0.5mg/mL
- Storage Buffer: Aqueous buffered solution containing ≤0.09% sodium azide
Specifications
Specifications
| Concentration | 0.5mg/mL |
| For Use With (Application) | ELISA, Flow Cytometry |
| Formulation | Aqueous Buffered solution containing ≤0.09% Sodium Azide |
| Name | Mouse CD1d:Ig Recombinant Fusion Protein |
| pH Range | 7.2 |
| Purification Method | Affinity Chromatography, SDS-PAGE |
| Quantity | 0.25 mg |
| Storage Requirements | 4°C |
| Cross Reactivity | Mouse |
| Recombinant | Recombinant |
| Show More |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction