missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD CD1d:Ig Recombinant Fusion Protein-Dimeric, Human
Click to view available options
Quantity:
0.1 μg
Unit Size:
Each
Description
- Format: Purified
- Concentration: 0.5mg/ml
- Isotype: Mouse IgG1, γ
- Reactivity: Human
- Storage buffer: Aqueous buffered solution containing ≤0.09% sodium azide
Flow Cytometry, Enzyme Linked Immunosorbent Assay
Specifications
Specifications
| Concentration | 0.5mg/mL |
| For Use With (Application) | ELISA, Flow Cytometry |
| Formulation | Aqueous Buffered solution containing ≤0.09% Sodium Azide |
| Name | Human CD1d:Ig Recombinant Fusion Protein |
| Purification Method | Affinity Chromatography, SDS-PAGE |
| Quantity | 0.1 μg |
| Storage Requirements | 4°C |
| Cross Reactivity | Human |
| Recombinant | Recombinant |
| Form | Solution |
Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction