Learn More
BD BrdU In-Situ Detection Kit IIrra
Description
Includes
Fixation Buffer, Diluent Buffer, BDRetrievagen A antigen retrieval solution, biotinylated anti-BrdU antibody, ready-to-use Streptavidin-Horseradish peroxidase (Sav-HRP) solution, DAB Buffer, DAB Chromogen, and Control Slides
The BrdU In-Situ Detection Kit features our BD Retrievagen A, an antigen retrieval solution designed specifically to unmask antigenic sites, preserve tissue morphology, and enable simultaneous staining of other surface antigens in conjunction with BrdU.
- With BD Retrievagen A, an antigen retrieval solution
- Designed specifically to unmask antigenic sites, preserve tissue morphology, and enable simultaneous staining of other surface antigens in conjunction with BrdU
- Kit enables study of proliferation state of phenotypically defined cells within micro-environment of tissues
- Consistent results are assured by providing all critical reagents
- Comprehensive series of protocols for staining BrdU in paraffin-embedded and frozen tissue sections and cultured cells, and in conjunction with other antigens
- Control slides are provided in the kit to serve as a reference
Immunohistochemical staining
Specifications
Specifications
| Quantity | 1 Kit |
| Storage Requirements | 4°C |
| For Use With (Application) | Immunohistochemical Staining |
Safety and Handling
- Danger
- Germ cell mutagenicity Category 2
- Carcinogenicity Category 1A
- Skin sensitiser Category 1
- Carcinogenicity Category 1B
- H317-May cause an allergic skin reaction.
- H341-Suspected of causing genetic defects.
- H350-May cause cancer.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P501-Dispose of contents/container to .
- P202-Do not handle until all safety precautions have been read and understood.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Research Use only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.