missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Taxo™ P Discs (Vial)

Product Code. 11702924
Change view
Click to view available options
Packaging Type:
6 Vials
Vial
Quantity:
50 Discs
6 x 50 Discs
Unit Size:
Pack of 300
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Type Quantity unitSize
11702924 Vial 50 Discs Pack of 50
11712924 6 Vials 6 x 50 Discs Pack of 300
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11702924 Supplier BD Supplier No. 231046

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For presumptive identification of pneumococci, and are intended for use with pure cultures

  • 6mm discs made from high quality absorbent paper
  • Impregnated with approximately 5.0μg of ethylhydrocupreine hydrochloride per disc

Order Info

Vials of 50 discs.

TRUSTED_SUSTAINABILITY

Specifications

Antimicrobial Agents Ethylhydrocupreine hydrochloride (optochin)
Concentration 5 μg
Content And Storage -20°C to +8°C
For Use With (Application) Differentiating Pneumococci
Packaging Type Vial
Quantity 50 Discs

For in vitro Diagnostic Use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.