Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Sulfamethoxazole-Trimethoprim
Product Code. 11757594
Change view
Click to view available options
Concentration:
23.75 μg/1.25 μg
Quantity:
10/Pk.
Unit Size:
Pack of 10
Product Code. Concentration Quantity unitSize
11757594 23.75 μg/1.25 μg 10/Pk. Pack of 10
1 options
Product Code. 11757594 Supplier BD Supplier No. 231539
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents
  • Clearly marked on both sides with letters and numbers designating agent and concentration
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Sulfamethoxazole, Trimethoprim
Concentration 23.75 μg/1.25 μg
Packaging Type Disc
Quantity 10/Pk.