Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Penicillin (10U)
Product Code. 11772954
Change view
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 500
Product Code. Quantity unitSize
11772954 10/Pk. Pack of 500
1 options
Product Code. 11772954 Supplier BD Supplier No. 231321
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents
  • Clearly marked on both sides with letters and numbers designating agent and concentration
  • Available in pack of 1 and 10
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Penicillin
Concentration 10U
Packaging Type Disc
Quantity 10/Pk.