Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Oxacillin
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
Description
- Discs containing certifiable agents are submitted to FDA for assay and certification before distribution Clearly marked on both sides with letters and numbers designating agent and concentration
Specifications
Specifications
| Antimicrobial Agents | Oxacillin |
| Concentration | 1 μg |
| Packaging Type | Disc |
| Quantity | 10/Pk. |