missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Nitrofurantoin (300μg)

Product Code. 17382371
Change view
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17382371 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17382371 Supplier BD Supplier No. 231293

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents

  • Discs containing certifiable agents are submitted to FDA for assay and certification before distribution Clearly marked on both sides with letters and numbers designating agent and concentration

TRUSTED_SUSTAINABILITY

Specifications

Antimicrobial Agents Nitrofurantoin
Concentration 300 μg
Packaging Type Disc
Quantity 10/Pk.
compliance-icons compliance-icons
Product Identifier
  • BD BBL Nitrofurantoin F/M-300 10 Ea
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 2
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 2
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P321-Specific treatment (see on this label).
  • P330-Rinse mouth.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Nitrofurantoin monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.