Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Minocyline
Product Code. 11783819
Change view
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 500
Product Code. Quantity unitSize
11783819 10/Pk. Pack of 500
1 options
Product Code. 11783819 Supplier BD Supplier No. 231251
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents
  • Discs containing certifiable agents are submitted to FDA for assay and certification before distribution Clearly marked on both sides with letters and numbers designating agent and concentration
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Minocycline
Concentration 30 μg
Packaging Type Disc
Quantity 10/Pk.