Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Disc Bacitracin, B-10
Product Code. 11792934
Change view
Click to view available options
Concentration:
10U
Unit Size:
Pack of 500
Product Code. Concentration unitSize
11792934 10U Pack of 500
1 options
Product Code. 11792934 Supplier BD Supplier No. 231268
Cost-effective method uses antimicrobial agents impregnated on high-quality paper
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents.
  • Employs Bauer-Kirby disc diffusion testing method
  • Use in self-tamping dispensers
  • Discs containing certifiable agents are submitted to FDA for assay and certification before distribution Clearly marked on both sides with letters and numbers designating agent and concentration
  • Cartridges of 50 discs each are sold individually or in packs of 10
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Bacitracin
Concentration 10U
Packaging Type Disc
Quantity 10 Cartridges