Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Disc Ampicillin with Sulbactam5, SAM-20
Click to view available options
Concentration:
10/10 μg
Unit Size:
Pack of 10
Description
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents.
- Employs Bauer-Kirby disc diffusion testing method
- Use in self-tamping dispensers
- Discs containing certifiable agents are submitted to FDA for assay and certification before distribution Clearly marked on both sides with letters and numbers designating agent and concentration
- Sold in packs of 10
Specifications
Specifications
| Antimicrobial Agents | Ampicillin with Sulbactam |
| Concentration | 10/10 μg |
| Packaging Type | Disc |
| Quantity | 10/Pk. |