Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Streptomycin
Product Code. 11737594
Click to view available options
Concentration:
300 μg
Unit Size:
Pack of 50
Product Code. 11737594
Brand: BD 231694
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
  • 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
  • Are clearly marked on both sides with letters and numbers designating agent and drug content
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Streptomycin
Concentration 300 μg
Packaging Type Disc
Quantity 1 Pack
For in vitro diagnostic use