Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Levofloxacin42
Product Code. 17392401
Change view
Click to view available options
Concentration:
5 μg
Unit Size:
Each
Product Code. Concentration unitSize
17392401 5 μg Each
1 options
Product Code. 17392401 Supplier BD Supplier No. 231705
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
  • 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
  • Are clearly marked on both sides with letters and numbers designating agent and drug content
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Levofloxacin
Concentration 5 μg
Packaging Type Disc
Quantity Each
For in vitro diagnostic use