missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Ceftazidime/Clavulanic Acid

Product Code. 17382441
Change view
Click to view available options
Quantity:
1 Pack
10 Pack
Unit Size:
Each
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17382441 1 Pack Each
17392441 10 Pack Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17382441 Supplier BD Supplier No. 231753

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens

  • 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
  • Are clearly marked on both sides with letters and numbers designating agent and drug content

TRUSTED_SUSTAINABILITY

Specifications

Antimicrobial Agents Ceftazidime, Clavulanic Acid
Concentration 10 μg
Packaging Type Disc
Quantity 1 Pack
compliance-icons
Product Identifier
  • Disc Ceftazidime/Clavulanic
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
Hazard Statement
  • H300-Fatal if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P321-Specific treatment (see on this label).
  • P330-Rinse mouth.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Ceftazidime

For in vitro diagnostic use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.