missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™/Difco™ Media Additives/Ingredients/Reagents: Saccharose

Product Code. 11748253
Click to view available options
:
Saccharose (Sucrose); BD Difco; 500g
This item is not returnable. View return policy

Product Code. 11748253

Brand: BD 217610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

D-saccharose, sucrose

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Content And Storage <30°C
Additive Saccharose
Form Powder
For Use With (Application) Dehydrated culture media
Packaging Type Bottle
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.