missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™/Difco™ Media Additives/Ingredients/Reagents: Penicillinase Concentrate

Product Code. 10615886
Click to view available options
Quantity:
20 mL
100 mL
Packaging Type:
Bottle
Tube
Unit Size:
200mL
Each
This item is not returnable. View return policy

Product Code. 10615886

Brand: BD 211898

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For cultivation of heterotrophic microorganisms

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Concentration 10000000U/mL
Additive Penicillinase
Form Liquid
For Use With (Application) Formulation of culture media for the cultivation of heterotrophic microorganisms
Packaging Type Tube
Quantity 20 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.