missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Dehydrated Culture Media: Phenol Red Dextrose Broth

Product Code. 12720187
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12720187 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12720187 Supplier BD Supplier No. 211514

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for determination of fermentation reactions in differentiation of microorganisms

Order Info

See our web site or the Fisher Scientific™ HealthCare catalog for product information.

Certifications

BAM

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance BAM
Form Dehydrated
Media Type Phenol Red Dextrose Broth
Product Type Microbiology Media
Formulation Pancreatic digest of Casein 10g/L, Sodium Chloride 5g/L, Phenol red 18mg/L, Dextrose 5g/L
For Use With (Application) Determination of fermentation reactions in the differentiation of microorganisms
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.