missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Dehydrated Culture Media: FTA Hemagglutination Buffer
Click to view available options
:
FTA Hemagglutination Buffer, pH 7.2 (Phosphate Buffered Saline); 500g
Description
- Phosphate Buffered Saline
- Formulation: Sodium Chloride 7.65g/L, Disodium Phosphate 1268.8mg/L, Monosodium Phosphate 0.1g/L, Monopotassium Phosphate 211.3mg/L
- pH 7.2
- Quantity: 500g
Specifications
Specifications
| Content And Storage | Refrigerate |
| Form | Dehydrated |
| Media Type | FTA Hemagglutination Buffer |
| Product Type | Buffer Solution |
| Formulation | Sodium Chloride 7.65g/L, Disodium Phosphate 1268.8mg/L, Monosodium Phosphate 0.1g/L, Monopotassium Phosphate 211.3mg/L |
| For Use With (Application) | Used for the FTA-ABS test and other serological procedures as a diluent and for washing slide preparations |
| Packaging Type | Bottle |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction