missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Dehydrated Culture Media: Eosin Methylene Blue Agar, Levine
Click to view available options
:
EMB Agar, Levine; 500g
Description
- Formulation: Pancreatic digest of Gelatin 10g/L, Lactose 10g/L, Dipotassium Phosphate 2g/L, Eosin Y 0.4g/L, Methylene blue 65mg/L, Agar 15g/L
Certifications
AOAC, BAM, BS12, CCAM, CMPH2, COMPF, MCM9, SMD
Specifications
Specifications
| Certifications/Compliance | AOAC, BAM, BS12, CCAM, CMPH2, COMPF, MCM9, SMD |
| Content And Storage | 2°C to 8°C |
| Form | Dehydrated |
| Media Type | EMB Agar, Levine |
| Product Type | Agar |
| Formulation | Pancreatic digest of Gelatin 10g/L, Lactose 10g/L, Dipotassium Phosphate 2g/L, Eosin Y 0.4g/L, Methylene blue 65mg/L, Agar 15g/L |
| For Use With (Application) | Isolation of gram negative enteric bacteria |
| Packaging Type | Bottle |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction