missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Dehydrated Culture Media: Dermatophyte Test Medium Base

Product Code. 11735055
Click to view available options
:
Dermatophyte Test Medium Base; 500g
This item is not returnable. View return policy

Product Code. 11735055

Brand: BD 212330

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For detection and presumptive identification of dermatophytes from clinical and veterinary specimens

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Refrigerate
Form Dehydrated
Media Type Dermatophyte Test Medium Base
Product Type Dehydrated Culture Media
Formulation Papaic digest of Soybean Meal 10g/L, Dextrose 10g/L, Phenol red 0.2g/L, Cycloheximide 0.5g/L, Agar 20g/L
For Use With (Application) Detection and presumptive identification of dermatophytes from clinical and veterinary specimens
Packaging Type Bottle
Quantity 500 g
compliance-icons compliance-icons
Product Identifier
  • Bottle Dermatophyte Test Med Base 500G
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H301-Toxic if swallowed.
  • H341-Suspected of causing genetic defects.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : protein hydrolyzates, Cicloheximide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.