missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Dehydrated Culture Media: Columbia Agar Base
Click to view available options
Quantity:
500 g
2.26 kg
Packaging Type:
Bottle
Pail
For Use With (Application):
Growth of fastidious microbes
Unit Size:
500g
Each
Specifications
Specifications
| Certifications/Compliance | AOAC, COMPF, EP, ISO, JP, USP |
| Content And Storage | 2°C to 8°C |
| Form | Dehydrated |
| Media Type | Columbia Agar Base |
| Product Type | Agar |
| Formulation | Pancreatic digest of Casein 10g/L, meat Peptic digest 5g/L, Yeast extract 5g/L, heart Pancreatic digest 3g/L, Corn Starch 1g/L, Sodium Chloride 5g/L, Agar 13.5g/L |
| For Use With (Application) | Growth of fastidious microbes |
| Packaging Type | Pail |
| Quantity | 2.26 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction