missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Dehydrated Culture Media: Columbia Agar Base

Product Code. 15142237
Click to view available options
Quantity:
500 g
2.26 kg
Packaging Type:
Bottle
Pail
For Use With (Application):
Growth of fastidious microbes
Unit Size:
500g
Each
This item is not returnable. View return policy

Product Code. 15142237

Brand: BD 211124

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for cultivation of fastidious microorganisms

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance AOAC, COMPF, EP, ISO, JP, USP
Content And Storage 2 to 25°C
Form Dehydrated
Media Type Columbia Agar Base
Product Type Agar
Formulation Pancreatic digest of Casein 10g/L, meat Peptic digest 5g/L, Yeast extract 5g/L, heart Pancreatic digest 3g/L, Corn Starch 1g/L, Sodium Chloride 5g/L, Agar 13.5g/L
For Use With (Application) Growth of fastidious microbes
Packaging Type Bottle
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.