missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Dehydrated Culture Media: Brucella Agar

Product Code. 11798103
Click to view available options
:
Brucella Agar; 500g
This item is not returnable. View return policy

Product Code. 11798103

Brand: BD 211086

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Dehydrated form for the convenience of the laboratorian and bioprocessing specialist

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CCAM, ISO, USDA
Form Dehydrated
Media Type Brucella Agar
Product Type Agar
Formulation Pancreatic digest of Casein 10g/L, Peptic digest of Animal Tissue 10g/L, Dextrose 1g/L, Yeast extract 2g/L, Sodium Chloride 5g/L, Sodium Bisulfite 0.1g/L, Agar 15g/L
For Use With (Application) Cultivation of Brucella organisms
Packaging Type Bottle
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.