missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Dehydrated Culture Media: BiGGY Agar
Click to view available options
:
BiGGY Agar; 500g
Description
- Formulation: Bismuth Ammonium Citrate 5g/L, Sodium Sulfite 3g/L, Dextrose 10g/L, Glycine 10g/L, Yeast extract 1g/L, Agar 16g/L
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Form | Dehydrated |
| Media Type | BIGGY Agar |
| Product Type | Agar |
| Formulation | Bismuth Ammonium Citrate 5g/L, Sodium Sulfite 3g/L, Dextrose 10g/L, Glycine 10g/L, Yeast extract 1g/L, Agar 16g/L |
| For Use With (Application) | For Candida |
| Packaging Type | Bottle |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction