missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Beef Extract Powder

Product Code. 16209781 Shop All Gibco Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16209781 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16209781 Supplier Gibco™ Supplier No. 212303
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Beef Extract Powder is a meat extract dried into a powder. This nutritional supplement provides a mixture of peptides, individual amino acids, nucleotides, organic acids, minerals, and vitamins.

Beef Extract Powder is a meat extract dried into a powder. This nutritional supplement provides a mixture of peptides, individual amino acids, nucleotides, organic acids, minerals, and vitamins. Additionally, due to differences in the process, Beef Extract Powder has a different molecular weight profile than other beef extract products in our portfolio. Customers have used Beef Extract Powder in a variety of microbial fermentation cell lines including Clostridium sp., Escherichia coli, Bacillus subtillis, Pasteurella multocida, and Streptococcus species to produce numerous animal health vaccines.

Features:

  • An animal origin (AO) nutritional supplement
  • A mixture of peptides, individual amino acids, nucleotides, organic acids, minerals, and vitamins

Applications of Beef Extract Powder include:

  • Used in a variety of microbial fermentation cell lines
  • Used to produce numerous animal health vaccines
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance AOAC, COMPF, SMWW, ISO/DIS, 11014
Content And Storage Store at 2°C to 25°C in cool, dry conditions in well-sealed containers.
Additive Beef Extract
Form Powder
For Use With (Application) Dehydrated culture media
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.