missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Yeast Extract

Product Code. 16229771 Shop All Gibco Products
Change view
Click to view available options
Quantity:
1 lb.
5 lb.
25 lb.
Unit Size:
11.30kg
2.2kg
454g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16229771 5 lb. 2.2kg
16219771 1 lb. 454g
16239771 25 lb. 11.30kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16229771 Supplier Gibco™ Supplier No. 211930
Make Your Process Development Count
Use promo code 27978
View all promotions
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Water-soluble extract of autolyzed yeast cells suitable for use in culture media.

Yeast Extracts are concentrates of the water-soluble portion of autolyzed Saccharomyces cerevisiae cells. They are derived from primary grown baker's yeast. These animal origin-free products are suitable for use as multi-functional nutritional supplements in mammalian cell culture, microbial fermentation, and insect cell culture applications. Yeast extract provides essential water-soluble vitamins, amino acids, peptides, and carbohydrates to any medium formulation.

Related yeast extract products:

  • Bacto Yeast Extract—one of the most complete and versatile fermentation supplements available. It is an important ingredient for the microbiological assay of vitamins. Yeast extract is also of value in the assay of antibiotics. B factor, a growth substance necessary for the production of rifampin in a Nocardia species, can be isolated from yeast extract.
  • Bacto Yeast Extract, Technical—developed to provide products priced for the biotechnology market with acceptable clarity and growth-promoting characteristics
  • Gibco Yeast Extract—developed to provide a product for the biotechnology/pharmaceutical market with acceptable clarity and growth-promoting characteristics. Media formulations containing yeast extract are specified in standard methods for various applications.
  • Difco Yeast Extract UF—ultra-filtered and specifically designed for mammalian cell culture applications. With its low endotoxin level and high content of naturally occurring B vitamins, it can be used effectively to reduce the need for fetal bovine serum. It has an endotoxin level of less than or equal to 500 EU/g.
  • Difco Yeast Extract Low-Dusting (LD)—designed to limit the amount of dust generated compared to other spray-dried yeast extracts.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance LUO, MUO
Content And Storage Store at 2°C to 25°C in cool, dry conditions in well-sealed containers.
Cell Type Bacteria
Culture Type Mammalian, Insect, Microbial
Form Powder
Product Type Bacterial Media
Format Pail
For Use With (Application) Formulation of culture media for the cultivation of heterotrophic microorganisms
Quantity 5 lb.
Source Animal origin-free
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.