missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Acidicase™ Peptone

Product Code. 16239761
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 16239761

Brand: Gibco™ 211843

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Applications:

Nutritional supplement in Vitamin assay, susceptibility testing and other laboratory media and microbial fermentation

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Content And Storage Storage conditions: 2°C to 25°C.  Store in cool, dry conditions in well-sealed containers.
Form Solid
Product Type Bacterial Media
Format Bottle
For Use With (Application) Nutritional supplement in Vitamin assay, susceptibility testing and other laboratory media and microbial fermentation
Product Line Acidicase
Quantity 500 g
Source Animal Origin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.