missing translation for 'onlineSavingsMsg'
Learn More

BD Bacto™ Dehydrated Culture Media: Todd Hewitt Broth

Product Code. p-4901289
Click to view available options
Quantity:
500 g
2 kg
10 kg
Packaging Type:
Bottle
Pail
For Use With (Application):
Hemolytic Streptococci growth
Form:
Dehydrated
Unit Size:
10kg
2kg
500g
This item is not returnable. View return policy

Product Code. 11708962

Brand: BD 249240

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For cultivation of group A streptococci, pathogenic microorganisms or as a blood culture medium

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Content And Storage <30°C
Form Dehydrated
Media Type Todd Hewitt Broth
Product Type Microbiology Media
Formulation Heart infusion from 500g 3.1g/L, Neopeptone 20g/L, Dextrose 2g/L, Sodium Chloride 2g/L, Disodium Phosphate 0.4g/L, Sodium Carbonate 2.5g/L
For Use With (Application) Hemolytic Streptococci growth
Packaging Type Bottle
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.