missing translation for 'onlineSavingsMsg'
Learn More

BD Bacto™ Dehydrated Culture Media: Todd Hewitt Broth

Product Code. 11708962
Change view
Click to view available options
Quantity:
500 g
2 kg
10 kg
Packaging Type:
Bottle
Pail
For Use With (Application):
Hemolytic Streptococci growth
Form:
Dehydrated
Unit Size:
10kg
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Type For Use With (Application) Form unitSize
11708962 500 g Bottle Hemolytic Streptococci growth Dehydrated 500g
11798952 10 kg Pail Hemolytic Streptococci growth Dehydrated 10kg
11788952 2 kg Pail Hemolytic Streptococci growth Dehydrated 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11708962 Supplier BD Supplier No. 249240

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For cultivation of group A streptococci, pathogenic microorganisms or as a blood culture medium

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Content And Storage <30°C
Form Dehydrated
Media Type Todd Hewitt Broth
Product Type Microbiology Media
Formulation Heart infusion from 500g 3.1g/L, Neopeptone 20g/L, Dextrose 2g/L, Sodium Chloride 2g/L, Disodium Phosphate 0.4g/L, Sodium Carbonate 2.5g/L
For Use With (Application) Hemolytic Streptococci growth
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.