missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Bacto™ Tryptone

Product Code. 16269751 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
2 kg
10 kg
Unit Size:
2kg
500g
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16269751 10 kg Each
16259751 2 kg 2kg
16279751 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16269751 Supplier Gibco™ Supplier No. 211701
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

An animal origin (AO) pancreatic digest of casein.

  • Bacto Tryptone is an animal origin (AO) pancreatic digest of casein
  • It is included in standard methods manual applications and is listed in the Reagent section of the United States Pharmacopeia as meeting the specifications for a pancreatic digest of casein, a component in many of the media listed
  • The nutritional profile of this supplement is unique in that it provides a range of nutrients
  • Customers have used this supplement in fermentation applications and shown that it successfully supports the growth of many bacterial species during the production of animal health vaccines
  • This includes Escherichia coli, and Fusobacterium and Agrobacterium species when used together with Bacto Yeast Extract
  • Store in cool, dry conditions in well-sealed containers at 2° to 25°C

Features:

  • Pancreatic digest of casein
  • Developed while investigating a peptone particularly suitable for the elaboration of indole by bacteria
  • Animal origin nutritional supplement

Applications of Bacto Tryptone include:

  • Works well in fermentation applications
  • Use with common organisms such as Escherichia coli, as well as uncommon organisms such as the diatom Nitzschia laevis
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance LUO, MUO
Content And Storage Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Form Powder
Product Type Bacterial Media
Format Pail
For Use With (Application) Dehydrated culture media
Product Line Bacto
Quantity 10 kg
Source Animal Origin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.