missing translation for 'onlineSavingsMsg'
Learn More

BD Mouse B Lymphocyte Activation Antibody Cocktail, with Isotype Control; PE-Cy™7 CD25, PE CD69, and FITC CD19

Brand:  BD 558064

Product Code. 15839188

  • ONLINE EXCLUSIVE PRICE
    £1000.00
    £957.54 / Each
    Save £42.46
    4% Off

Please to purchase this item. Need a web account? Register with us today!


Includes: Mouse B Lymphocyte Activation Antibody Cocktail; PE-Cy™7 CD25, PE CD69, and APC CD19, Mouse B Lymphocyte Activation Isotype Control; PE-Cy 7, PE, and APC
This item is not returnable. View return policy

Description

Description

Three antibodies have been titrated and pre-diluted, mixed together, and formulated for optimal staining performance. The Mouse B Lymphocyte Activation Isotype Control contains equivalent concentrations of fluorochrome- and isotype-matched negative-control immunoglobulin

  • Clone: 1D3, H1.2F3, PC61
  • Isotype: Rat IgG2a, κ, Armenian Hamster IgG1, λ3, Rat IgG1,λ
  • Permits recognition of each of the three antigens on each cell in sample
  • Expression of the three antigens identifies activated B lymphocytes
  • Additional fluorochrome-labeled reagents may be combined
  • Monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography

Flow Cytometry

TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 Set
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Research use only