missing translation for 'onlineSavingsMsg'
Learn More

BD Annexin V-PE

Product Code. 10092374
Change view
Click to view available options
Quantity:
100 Tests
200 Tests
25 Tests
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10092374 200 Tests Each
12860460 100 Tests Each
15840609 25 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10092374 Supplier BD Supplier No. 556421

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Serves as a sensitive probe for flow cytometric analysis of cells that are undergoing apoptosis

  • Format: PE

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Quantity 200 Tests
Sample Volume 5 μL
Storage Buffer Aqueous Buffered solution containing BSA and ≤0.09% Sodium Azide
Content And Storage 4°C
For Use With (Application) Flow Cytometry
Form Liquid

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.