missing translation for 'onlineSavingsMsg'
Learn More

BD Annexin V: FITC Fluorescence Microscopy Kit

Product Code. 15846428 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
25 Slides
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15846428 25 Slides 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15846428 Supplier BD Supplier No. 550911

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Annexin V-FITC, Annexin V Binding Buffer, and PBS Buffer

Fluorescence staining for apoptosis

Early in apoptosis, before the loss of membrane integrity,phosphatidylserine (PS) translocates from the inner to outer leaflet of plasma membrane, exposing it to external cellular environment at surface of plasma membrane. Annexin V is 35-36kDa, calcium-dependent, phospholipid-binding protein with a high affinity for PS and is used to indirectly monitor PS translocation. Annexin V assay is facilitated by the observation that PS translocation appears to be a universal apoptotic phenomenon.

  • Binds to negatively charged phospholipid surfaces
  • Defined calcium and salt concentrations are required

Used in Fluorescence microscopy

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Fluorescence Microscopy
Includes Annexin V-FITC, Annexin V binding buffer, and PBS buffer
Quantity 25 Slides
Content And Storage 4°C
Product Type Fluorescence Microscopy Kit

Research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.