missing translation for 'onlineSavingsMsg'
Learn More

BD Akt/PKB Sampler Kit

For intracellular transduction of insulin-induced signals and anti-apoptotic effects of multiple survival factors

Brand:  BD 611437

Product Code. 15855929

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Includes: Bad, GSK-3b, Insulin Receptor b, IRS-1, p70s6k, PI3-Kinase, PKBa/Akt, PKB Kinase/PDK1, PP2A Catalytic a
This item is not returnable. View return policy

Description

Description

  • Format: Purified
  • Storage buffer: Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide

Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Research Use Only