Learn More
BD AEC Substrate Kit
Substrate for peroxidase and is a widely used chromogen for immunohistochemical staining and immunoblotting
Brand: BD 551951
Additional Details : Weight : 0.11000kg Transport : UN number : 2265 Chem class : 3 Pack group : III
Alert:
Contains Dimethylsulfoxide, Methanol, Urea Peroxide, 3-Amino-9-Ethylcarbazole, and N, N,Dimethylformamide which are considered as harmful.
Includes: AEC Buffer and AEC Chromogen
Description
Immunohistochemistry, ELISPOT
Specifications
3mL | |
Immunohistochemical staining and immunoblotting, ELISPOT | |
10 plates, AEC buffer and AEC chromogen | |
4°C |
AEC | |
AEC Substrate | |
7.2 | |
100mL |
Safety and Handling
- BD ELISPOT AEC Substrate Set
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Reproductive toxicity Category 1B
- Specific target organ toxicity Category 2
Hazard Statement
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H350-May cause cancer.
- H360D-May damage the unborn child.
- H371-May cause damage to organs.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P330-Rinse mouth.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P370+P378-In case of fire: Use to extinguish.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : diemthyl formamide,3-amino-9-ethyl carbazole
Research Use Only