missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Ac-IETD-CHO Caspase-8 Inhibitor
Click to view available options
Quantity:
1 mg
Unit Size:
Each
Description
Members of caspase family have key roles in inflammation and mammalian apoptosis. Caspase-8 (FLICE/MACH-1) is a 55 kDa cytosolicprotein with homology to the CD95/Fas-associated signal transduction molecule, FADD, in addition to its homology with other caspases. Caspase-8 is activated early in apoptosis and is involved in the proteolysis and activation of pro-caspase-3
- Synthetic tetrapeptide inhibitor of caspase-8
- Contains the amino acid sequence that is the target for caspase-8 mediated proteolysis
- Used as a negative (blocking) control
- May be used in protease assays
- Format: Purified
Fluorescence quantitation
Specifications
Specifications
| For Use With (Application) | Fluorescence quantitation |
| Storage Buffer | Lyophilized in Dimethyl Sulfoxide |
| Content And Storage | -20°C |
| Quantity | 1 mg |
| Molecular Weight (g/mol) | 55 |
| Inhibitors | Ac-IETD-CHO |
| Form | Purified |
Research use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction