missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD A431 + EGF Cell Lysate
Click to view available options
Quantity:
500 μg
Unit Size:
Each
Description
Includes
Human Epidermoid Carcinoma
A source that can be rich in endogenously expressed proteins, this lysate is convenient to use and provided in a denatured format allowing for immediate loading onto an SDS-PAGE gel. A431 cells (Human epithelial carcinoma; ATCC CRL-1555) were grown to 80-90% confluency, serum starved overnight, then treated with 100ng/mL EGF for 5 minutes at 37°C.
- Storage buffer: SDS-PAGE buffer (62mM Tris pH 6.8, 2% SDS, 0.9% b-mercaptoethanol, 0.003% bromophenol blue, 5% glycerol)
Western blot
Specifications
Specifications
| For Use With (Application) | Western Blot |
| Tissue | Epidermoid Carcinoma |
| Quantity | 500 μg |
| Storage Buffer | 1mg/mL in SDS-PAGE Buffer: 62mM Tris pH 6.8, 2% SDS, 0.9% beta-mercaptoethanol, 0.003% Bromophenol Blue, 5% Glycerol |
| Host Species | Human |
| Concentration | 1 mg/mL |
| Content And Storage | -20°C |
Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction