missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ BD 1047 dihydrobromide
GREENER_CHOICE

Product Code. 30001398
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001398

Brand: Tocris Bioscience™ 0956/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

σ1 selective antagonist

σ1 receptor antagonist, exhibiting a similar binding profile to that of BD 1063 (Cat. No. 0883) but with higher affinity (approximately 10-fold) at both σ1 and σ2 sites.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138356-20-4
Molecular Formula C13H20Cl2N2

Specifications

Quantity 10 mg
Formula Weight 437.04
Purity >99%
Molecular Weight (g/mol) 275.217
Chemical Name or Material BD 1047 dihydrobromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.