missing translation for 'onlineSavingsMsg'
Learn More

BCX 1470 methanesulfonate, MedChemExpress

Product Code. 16408128
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16408128

Brand: Medchem Express HY50875

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BCX 1470 methanesulfonate inhibits the esterolytic activity of factor D (IC50=96 nM) and C1s (IC50=1.6 nM), 3.4- and 200-fold better, respectively, than that of trypsin.

BCX 1470 methanesulfonate inhibits the esterolytic activity of factor D (IC50=96 nM) and C1s (IC50=1.6 nM), 3.4- and 200-fold better, respectively, than that of trypsin.

Specifications

Chemical Name or Material BCX 1470 methanesulfonate
CAS 217099-44-0
Color White
Purity Grade Notes Research
Molecular Formula C15H14N2O5S3
Quantity 10 mg
Solubility Information DMSO : ≥ 33.33 mg/mL (83.64 mM)
SMILES N=C(C1=CC2=CC=C(OC(C3=CC=CS3)=O)C=C2S1)N.O=S(O)(C)=O
Molecular Weight (g/mol) 398.48
Formula Weight 398.48
Percent Purity 98.17%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.