missing translation for 'onlineSavingsMsg'
Learn More

BCS1L Antibody (5F3), Novus Biologicals™

Product Code. 18341538 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18341538 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18341538 Supplier Novus Biologicals Supplier No. H00000617M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

BCS1L Monoclonal antibody specifically detects BCS1L in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen BCS1L
Applications Western Blot, ELISA
Classification Monoclonal
Clone 5F3
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_004319
Gene Alias BCS, BCS1, BCS1 (yeast homolog)-like, BCS1-like (S. cerevisiae), BCS1-like (yeast), BCS1-like protein, BJSGRACILE, FLNMS, h-BCS, h-BCS1, mitochondrial chaperone BCS1, mitochondrial complex III assembly, PTD
Host Species Mouse
Immunogen BCS1L (NP_004319, 320 a.a. ~ 418 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. ASTEARIVFMTTNHVDRLDPALIRPGRVDLKEYVGYCSHWQLTQMFQRFYPGQAPSLAENFAEHVLRATNQISPAQVQGYFMLYKNDPVGAIHNAESLR
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 617
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.