missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCR Mouse anti-Human, Clone:6B5E12, Abnova™
Mouse monoclonal antibody raised against partial recombinant human BCR.
Brand: Abnova MAB17934.100ug
This item is not returnable.
View return policy
Description
A reciprocal translocation between chromosomes 22 and 9 produces the Philadelphia chromosome, which is often found in patients with chronic myelogenous leukemia. The chromosome 22 breakpoint for this translocation is located within the BCR gene. The translocation produces a fusion protein which is encoded by sequence from both BCR and ABL, the gene at the chromosome 9 breakpoint. Although the BCR-ABL fusion protein has been extensively studied, the function of the normal BCR gene product is not clear. The protein has serine/threonine kinase activity and is a GTPase-activating protein for p21rac. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]Specifications
| BCR | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:100-1:500) The optimal working dilution should be determined by the end user. |
|
| BCR | |
| Mouse | |
| 100 μg | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Western Blot | |
| 6B5E12 | |
| breakpoint cluster region | |
| In PBS (0.05% sodium azide) | |
| ALL, BCR-ABL1, BCR1, CML, D22S11, D22S662, FLJ16453, PHL | |
| Recombinant protein corresponding to amino acids 139-280 of human BCR. | |
| 613 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction