missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCR Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-19098AF488
This item is not returnable.
View return policy
Description
BCR Polyclonal antibody specifically detects BCR in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence
Specifications
| BCR | |
| Polyclonal | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| ALLD22S11, BCR1, breakpoint cluster region, breakpoint cluster region protein, CMLEC 2.7.11.1, D22S662, PHLFLJ16453, Renal carcinoma antigen NY-REN-26 | |
| The immunogen recognized by this antibody maps to a region between residue 1221 and 1271 of human breakpoint cluster region using the numbering given in entry NP_004318.3 (GeneID 613). | |
| 0.1 mL | |
| Phospho Specific, Protein Kinase, Signal Transduction | |
| 613 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Flow Cytometry, Immunofluorescence | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction