missing translation for 'onlineSavingsMsg'
Learn More

BCR Antibody, Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18417763 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18417763 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18417763 Supplier Novus Biologicals Supplier No. NBP119098AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

BCR Polyclonal antibody specifically detects BCR in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen BCR
Applications Western Blot, Flow Cytometry, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias ALLD22S11, BCR1, breakpoint cluster region, breakpoint cluster region protein, CMLEC 2.7.11.1, D22S662, PHLFLJ16453, Renal carcinoma antigen NY-REN-26
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1221 and 1271 of human breakpoint cluster region using the numbering given in entry NP_004318.3 (GeneID 613).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 613
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.