missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BCOR Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106545
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total BCOR.
Transcriptional corepressor. May specifically inhibit gene expression when recruited to promoter regions by sequence-specific DNA-binding proteins such as BCL6 and MLLT3. This repression may be mediated at least in part by histone deacetylase activities which can associate with this corepressor. Involved in the repression of TFAP2A; impairs binding of BCL6 and KDM2B to TFAP2A promoter regions. Via repression of TFAP2A acts as a negative regulator of osteo-dentiogenic capacity in adult stem cells; the function implies inhibition of methylation on histone H3 'Lys-4' (H3K4me3) and 'Lys-36' (H3K36me2).
Specifications
| BCOR | |
| Polyclonal | |
| Unconjugated | |
| Bcor | |
| 5830466J11Rik; 8430401K06Rik; ANOP2; BCL6 corepressor; BCL-6 corepressor; BCL6 co-repressor; BCL6 interacting corepressor; BCL-6 interacting corepressor; Bcor; BcorR; D930024N20Rik; Kiaa1575; MAA2; MCOPS2; mKIAA1575; RGD1562735 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 54880, 71458 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6W2J9, Q8CGN4 | |
| Bcor | |
| A synthesized peptide derived from human BCOR(Accession Q6W2J9), corresponding to amino acid residues V1237-F1287. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction