Learn More
BCOR Mouse, anti-Human, Clone: 1F12H6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68611-1-IG-150UL
This item is not returnable.
View return policy
Description
The BCOR antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human BCOR. This antibody recognizes human, mouse antigen. The BCOR antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| BCOR | |
| 1F12H6 | |
| ANOP2, BCOR, FLJ20285, FLJ38041, KIAA1575, MAA2, MCOPS2 | |
| 150 μL | |
| 54880 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG1 |