missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BCMO1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5122212
This item is not returnable.
View return policy
Description
Vitamin A metabolism is important for vital processes such as vision, embryonic development, cell differentiation, and membrane and skin protection. The protein encoded by this gene is a key enzyme in beta-carotene metabolism to vitamin A. It catalyzes the oxidative cleavage of beta, beta-carotene into two retinal molecules.
Specifications
| BCMO1 | |
| Polyclonal | |
| Unconjugated | |
| BCO1 | |
| BCDO; BCDO1; BCMO; Bcmo1; BCO; BCO1; Beta,beta-carotene 15,15'-dioxygenase; beta,beta-carotene 15,15'-monooxygenase; beta-carotene 15 15'-dioxygenase; beta-carotene 15 15-dioxygenase; beta-carotene 15, 15'-dioxygenase; beta-carotene 15, 15-dioxygenase; beta-carotene 15, 15'-dioxygenase 1; beta-carotene 15,15' dioxygenase; beta-carotene 15,15'-monooxygenase; beta-carotene 15,15'-monooxygenase 1; beta-carotene cleavage enzyme; beta-carotene dioxygenase 1; Beta-carotene oxygenase 1; beta-CD; betaCMOOX; carotenoid-15,15'-oxygenase; CMO1; Cmoi | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 100192320, 114106, 53630 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q91XT5, Q9HAY6 | |
| BCO1 | |
| Gly6-Pro206 (Accession Q9HAY6), with N-terminal His Tag. | |
| 100 μL | |
| Primary | |
| Human, Pig, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction