missing translation for 'onlineSavingsMsg'
Learn More

BCMA/TNFRSF17 Antibody (Vicky-1) - BSA Free, Novus Biologicals™

Product Code. 18485691 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
100 μg
Unit Size:
0.05mg
100µg
25µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18485691 100 μg 100µg
18031067 0.025 mg 25µg
18002157 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18485691 Supplier Novus Biologicals Supplier No. NBP197637100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 4 publications

BCMA/TNFRSF17 Monoclonal specifically detects BCMA/TNFRSF17 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BCMA/TNFRSF17
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Blocking Assay
Classification Monoclonal
Clone Vicky-1
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Blocking/Neutralizing 50-200 molar excess
Formulation PBS with 0.02% Sodium Azide
Gene Alias B cell maturation antigen, B-cell maturation protein, BCMAtumor necrosis factor receptor superfamily member 17, BCMB-cell maturation factor, CD269, CD269 antigen, tumor necrosis factor receptor superfamily, member 17
Gene Symbols TNFRSF17
Host Species Rat
Immunogen Recombinant human BCMA extracellular domain (aa 2-54).
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 608
Test Specificity BCMA. Does not cross-react with Mouse BCMA.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.